Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253702.1 | NP_001240631.1 | 2 | 817 | |
NM_001253703.1 | NP_001240632.1 | 2 | 817 | ||
NM_153091.3 | NP_694731.1 | 2 | 817 | ||
GenBank mRNA | AB084518.1 | - | 2 | 231 | |
AK033865.1 | - | 2 | 249 | ||
AK144075.1 | - | 2 | 257 | ||
AK148099.1 | - | 2 | 519 | ||
AK172102.1 | - | 2 | 818 | ||
AK172218.1 | - | 2 | 818 | ||
BC024473.1 | - | 2 | 208 | ||
BC058781.1 | - | 2 | 242 | 6311899 | |
BC107394.2 | - | 2 | 242 | 40058131 | |
BC145950.1 | - | 2 | 309 | 40131253 |