Gene Transcripts
- Gene Symbol
- SLC2A4
- Entrez Gene ID
- 6517
- Gene Name
- solute carrier family 2 member 4
- Gene Aliases
- GLUT4
- Chromosome Location
- Chr. 17: 7281735 - 7288048 on Build GRCh38
- UniGene ID
- Hs.380691
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042.2 |
NP_001033.1
|
3, 4 |
531 |
|
GenBank mRNA |
AK300899.1 |
- |
3, 4 |
598 |
|
AK313372.1 |
- |
3, 4 |
545 |
|
AL521328.3 |
- |
3, 4 |
530 |
|
BC034387.1 |
- |
3, 4 |
531 |
5187454 |
BC069615.1 |
- |
3, 4 |
426 |
7262457 |
BC069621.1 |
- |
3, 4 |
426 |
7262469 |
BC113592.1 |
- |
3, 4 |
358 |
8322644 |
BC126164.1 |
- |
3, 4 |
358 |
8991880 |
M20747.1 |
- |
3, 4 |
476 |
|
View Full Product Details