Gene Transcripts

Gene Symbol
Ttc39a
Entrez Gene ID
230603
Gene Name
tetratricopeptide repeat domain 39A
Gene Aliases
4922503N01Rik
Chromosome Location
Chr. 4: 109394148 - 109444745 on Build GRCm38
UniGene ID
Mm.33091
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145948.1 NP_001139420.1 11, 12 945
NM_153392.2 NP_700441.2 11, 12 954
GenBank mRNA AK048659.1 - 11, 12 955
AK076641.1 - 11, 12 907
AK076720.1 - 11, 12 946
AK076772.1 - 11, 12 908
AK172951.1 - 11, 12 877
BC023883.1 - 11, 12 953 5150841
BC026660.1 - 11, 12 895 4988900

View Full Product Details