Gene Transcripts

Gene Symbol
Rell2
Entrez Gene ID
225392
Gene Name
RELT-like 2
Gene Aliases
4631403P03Rik, ependolin
Chromosome Location
Chr. 18: 37955084 - 37959179 on Build GRCm38
UniGene ID
Mm.233516
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153793.2 NP_722488.2 4 676
GenBank mRNA AK044097.1 - 4 639
BC038500.1 - 4 619 4481989

View Full Product Details