Gene Transcripts

Gene Symbol
Fam13a
Entrez Gene ID
58909
Gene Name
family with sequence similarity 13, member A
Gene Aliases
AI462012, D430015B01Rik, FAM13A1, Precm1
Chromosome Location
Chr. 6: 58933536 - 59024502 on Build GRCm38
UniGene ID
Mm.28813
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153574.2 NP_705802.1 18 2432
GenBank mRNA AJ510136.1 - 18 2418
AK036291.1 - 11 1639
AK052421.1 - 18 2373
AK087913.1 - 646
BC080861.1 - 18 2432 30357292

View Full Product Details