Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153577.2 | NP_705805.1 | 5 | 923 | |
GenBank mRNA | AA407649.1 | - | 3 | 394 | |
BC004761.1 | - | 5 | 872 | 3155758 | |
BC023803.1 | - | 5 | 910 | 5352953 | |
BC056649.1 | - | 5 | 862 | 5036575 |