Gene Transcripts

Gene Symbol
Igsf11
Entrez Gene ID
207683
Gene Name
immunoglobulin superfamily, member 11
Gene Aliases
1700025L02Rik, BT-IgSF, Igsf13
Chromosome Location
Chr. 16: 38892476 - 39027159 on Build GRCm38
UniGene ID
Mm.245564
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170599.2 NP_733548.2 6 1002
GenBank mRNA AB079880.1 - 6 839
AK160146.1 - 6 965
BC057555.1 - 6 1002 6416190

View Full Product Details