Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172477.4 | NP_766065.1 | 2 | 569 | |
GenBank mRNA | AK047138.1 | - | 3 | 557 | |
AK081176.1 | - | 2 | 570 | ||
AK160784.1 | - | 2 | 331 | ||
AK165202.1 | - | 2 | 425 | ||
AK167298.1 | - | 2 | 423 | ||
BC056367.1 | - | 2 | 448 | ||
BC057310.1 | - | 2 | 448 | ||
BC145599.1 | - | 1 | 251 | 9054165 | |
BC150799.1 | - | 1 | 251 | 9087710 |