Gene Transcripts

Gene Symbol
Zfp11
Entrez Gene ID
22648
Gene Name
zinc finger protein 11
Gene Aliases
6720465D24, Krox-6, Zfp-11
Chromosome Location
Chr. 5: 129654595 - 129670088 on Build GRCm38
UniGene ID
Mm.135067
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172462.4 NP_766050.1 3 2144
GenBank mRNA AK032872.1 - 3 2159
AK085077.1 - 2 2130
BC058553.1 - 3 2142
BC094590.1 - 2 2129 30616053
BC139312.1 - 2 2124 8862334
BC139315.1 - 2 2124 8862337

View Full Product Details