Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163.3 | NP_001154.2 | 2 | 1085 | |
XM_005251968.3 | XP_005252025.1 | 2 | 1116 | ||
XM_011518617.2 | XP_011516919.1 | 2 | 1040 | ||
XM_017014670.1 | XP_016870159.1 | 2 | 1329 | ||
GenBank mRNA | AF029106.1 | - | 2 | 1007 | |
AF047347.1 | - | 2 | 1048 | ||
L04953.1 | - | 1 | 422 |