Gene Transcripts

Gene Symbol
Snx14
Entrez Gene ID
244962
Gene Name
sorting nexin 14
Gene Aliases
B830022K16, C330035N22Rik, YR-14
Chromosome Location
Chr. 9: 88376747 - 88439565 on Build GRCm38
UniGene ID
Mm.439873
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172926.3 NP_766514.2 3 444
GenBank mRNA AK046839.1 - 3 386
AK049371.1 - 3 423
AK082389.1 - 3 423
BC080799.1 - 3 429 4948538

View Full Product Details