Gene Transcripts

Gene Symbol
Pp2d1
Entrez Gene ID
110332
Gene Name
protein phosphatase 2C-like domain containing 1
Gene Aliases
4921523A10Rik
Chromosome Location
Chr. 17: 53507460 - 53539451 on Build GRCm38
UniGene ID
Mm.444633
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173449.3 NP_775625.1 2 371
GenBank mRNA AK076579.1 - 2 372

View Full Product Details