Gene Transcripts

Gene Symbol
Rbmx2
Entrez Gene ID
209003
Gene Name
RNA binding motif protein, X-linked 2
Gene Aliases
2700089A17Rik, 2810411A03Rik, CGI-79
Chromosome Location
Chr. X: 48695004 - 48710719 on Build GRCm38
UniGene ID
Mm.32561
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173376.3 NP_775552.1 2, 3 244
GenBank mRNA AK076115.1 - 2, 3 245
AK160232.1 - 2, 3 172
AK168018.1 - 2, 3 159
BC026976.1 - 2, 3 169 5361950

View Full Product Details