Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001218.4 | NP_001209.1 | 7 | 1039 | |
NM_001293642.1 | NP_001280571.1 | 6 | 859 | ||
NM_206925.2 | NP_996808.1 | 7 | 1039 | ||
NR_135511.1 | 7 | 1142 | |||
GenBank mRNA | AF037335.1 | - | 7 | 764 | |
AF051882.1 | - | 7 | 656 | ||
AK096845.1 | - | 6 | 595 | ||
AK315769.1 | - | 7 | 805 | ||
BC000278.1 | - | 7 | 761 | 3357439 | |
BC011691.2 | - | 7 | 764 | 3344734 | |
BC023981.1 | - | 7 | 779 | 4042589 | |
BT006656.1 | - | 7 | 649 |