Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173762.4 | NP_776123.3 | 35, 36 | 5543 | |
GenBank mRNA | AK049676.1 | - | 2, 3 | 345 | |
AY493378.1 | - | 35, 36 | 5456 | ||
BC049989.1 | - | 1, 2 | 108 | ||
BC059032.1 | - | 3, 4 | 505 |