Gene Transcripts
- Gene Symbol
- Synpo2l
- Entrez Gene ID
- 68760
- Gene Name
- synaptopodin 2-like
- Gene Aliases
- 1110054M18Rik, Chap
- Chromosome Location
- Chr. 14: 20658945 - 20668349 on Build GRCm38
- UniGene ID
- Mm.35789
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175132.3 |
NP_780341.1
|
1, 2 |
202 |
|
GenBank mRNA |
AK053054.1 |
- |
1, 2 |
202 |
|
View Full Product Details