Gene Transcripts

Gene Symbol
Ankrd9
Entrez Gene ID
74251
Gene Name
ankyrin repeat domain 9
Gene Aliases
2500003O20Rik
Chromosome Location
Chr. 12: 110975353 - 110979021 on Build GRCm38
UniGene ID
Mm.250989
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175207.4 NP_780416.2 3 2026
GenBank mRNA AK032641.1 - 3 2027
AK051108.1 - 3 2046

View Full Product Details