Gene Transcripts

Gene Symbol
AFG3L1P
Entrez Gene ID
172
Gene Name
AFG3 like matrix AAA peptidase subunit 1, pseudogene
Gene Aliases
AFG3, AFG3L1
Chromosome Location
Chr. 16: 89972580 - 90000787 on Build GRCh38
UniGene ID
Hs.534773
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003226.1 11 2802
NR_003227.1 10 2563
GenBank mRNA AK056488.1 - 12 2408
AK090856.1 - 2 2013
AK091293.1 - 11 2765
AK097044.1 - 10 2550
AK126911.1 - 9 2593
AK128352.1 - 4 2402
AK128804.1 - 5 1929
AL832984.1 - 5 2026
BC021245.2 - 9 2375

View Full Product Details