Gene Transcripts

Gene Symbol
Six3os1
Entrez Gene ID
100043902
Gene Name
SIX homeobox 3, opposite strand 1
Gene Aliases
D17Mgi26, E130112H22Rik, Rncr1, Six3os
Chromosome Location
Chr. 17: 85601932 - 85618396 on Build GRCm38
UniGene ID
Mm.150838
Species
Mus musculus
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK044330.1 - LIT3371
RNCR1
2 960

View Full Product Details