Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175414.4 | NP_780623.1 | 4 | 305 | |
GenBank mRNA | AK079139.1 | - | 4 | 278 | |
AK158729.1 | - | 3 | 190 | ||
AK163395.1 | - | 4 | 361 | ||
AK189318.1 | - | 162 | |||
AK199601.1 | - | 2 | 111 | ||
BC052503.1 | - | 4 | 287 | 5146854 | |
BY030381.1 | - | 4 | 305 |