Gene Transcripts

Gene Symbol
Mrgprb2
Entrez Gene ID
243979
Gene Name
MAS-related GPR, member B2
Gene Aliases
4833406I20Rik, Mgrg14
Chromosome Location
Chr. 7: 48550965 - 48658094 on Build GRCm38
UniGene ID
Mm.83689
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175531.4 NP_780740.2 2 181
GenBank mRNA AK029369.1 - 2 182
BC107390.1 - 2 179 40057815
BC107391.2 - 2 179 40057816

View Full Product Details