Gene Transcripts

Gene Symbol
Ankar
Entrez Gene ID
319695
Gene Name
ankyrin and armadillo repeat containing
Gene Aliases
4932422E22Rik
Chromosome Location
Chr. 1: 72642980 - 72700579 on Build GRCm38
UniGene ID
Mm.249056
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176980.4 NP_795954.2 2 330
GenBank mRNA AK030065.1 - 2 331
BC132415.1 - 2 305 40130692
BC145253.1 - 2 305 9053805

View Full Product Details