Gene Transcripts

Gene Symbol
Nwd1
Entrez Gene ID
319555
Gene Name
NACHT and WD repeat domain containing 1
Gene Aliases
A230063L24Rik
Chromosome Location
Chr. 8: 72646225 - 72716880 on Build GRCm38
UniGene ID
Mm.212927
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176940.5 NP_795914.3 5 1080
GenBank mRNA AK034868.1 - 779
AK038792.1 - 4 772
AK162888.1 - 5 1075
BC082552.1 - 5 1020
BC144786.1 - 3 670 9053334
BC144787.1 - 3 670
BC145703.1 - 3 670 40131006

View Full Product Details