Gene Transcripts

Gene Symbol
Rimkla
Entrez Gene ID
194237
Gene Name
ribosomal modification protein rimK-like family member A
Gene Aliases
B930030J24, NAAGS, NAAGS-II, Rimk
Chromosome Location
Chr. 4: 119465279 - 119493153 on Build GRCm38
UniGene ID
Mm.206201
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177572.4 NP_808240.2 5 1002
GenBank mRNA AK047164.1 - 5 997
AK163360.1 - 5 995
BC057371.1 - 5 983 6835897

View Full Product Details