Gene Transcripts

Gene Symbol
Ttc22
Entrez Gene ID
230576
Gene Name
tetratricopeptide repeat domain 22
Gene Aliases
4732467L16Rik
Chromosome Location
Chr. 4: 106622449 - 106640196 on Build GRCm38
UniGene ID
Mm.41945
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177667.4 NP_808335.1 7 2200
GenBank mRNA AK028895.1 - 7 2201
BC096484.1 - 7 2177

View Full Product Details