Gene Transcripts

Gene Symbol
Tgm6
Entrez Gene ID
241636
Gene Name
transglutaminase 6
Gene Aliases
TGM3L, TGY
Chromosome Location
Chr. 2: 130112416 - 130154232 on Build GRCm38
UniGene ID
Mm.83437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289747.1 NP_001276676.1 4 509
NM_001289748.1 NP_001276677.1 3 346
NM_177726.4 NP_808394.1 3 365
GenBank mRNA AK037229.1 - 3 355
AY159126.1 - 3 355
AY177607.1 - 3 346
BC112420.1 - 3 342 40061385
BC113796.1 - 4 486 40061392

View Full Product Details