Gene Transcripts

Gene Symbol
Aknad1
Entrez Gene ID
329738
Gene Name
AKNA domain containing 1
Gene Aliases
4921525H12Rik
Chromosome Location
Chr. 3: 108739560 - 108782309 on Build GRCm38
UniGene ID
Mm.72322
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177859.3 NP_808527.2 2 704
GenBank mRNA AK029552.1 - 2 582
AK164420.1 - 2 705

View Full Product Details