Gene Transcripts

Gene Symbol
Oaf
Entrez Gene ID
102644
Gene Name
OAF homolog (Drosophila)
Gene Aliases
AL024221, D130038B21Rik, D9Ucla1
Chromosome Location
Chr. 9: 43221278 - 43239816 on Build GRCm38
UniGene ID
Mm.246479
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178644.3 NP_848759.1 4 1907
GenBank mRNA AK051349.1 - 4 1907

View Full Product Details