Gene Transcripts

Gene Symbol
Mfsd11
Entrez Gene ID
69900
Gene Name
major facilitator superfamily domain containing 11
Gene Aliases
2600014M03Rik
Chromosome Location
Chr. 11: 116852283 - 116875660 on Build GRCm38
UniGene ID
Mm.348004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178620.3 NP_848735.1 13 2189
GenBank mRNA AK031384.1 - 13 2384
AK032043.1 - 13 2385
BC079583.1 - 13 2216
BC088988.1 - 13 2189 6418757

View Full Product Details