Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATCHX1
Entrez Gene ID
838208
Gene Name
Cation/hydrogen exchanger family protein
Gene Aliases
AT1G16380, CATION EXCHANGER 1, CHX1, F3O9.18, F3O9_18
Chromosome Location
Chr.1: 5598411 - 5601367 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101504.2 NP_173088.1 1 - 2 161 127
GenBank mRNA AY926465.1 - 1 - 2 161 127

View Full Product Details