Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RDH2
Entrez Gene ID
838216
Gene Name
rhodanese homologue 2
Gene Aliases
ARABIDOPSIS THALIANA MERCAPTOPYRUVATE SULFURTRANSFERASE 2, AT1G16460, ATMST2, ATRDH2, F3O9.26, F3O9_26, MERCAPTOPYRUVATE SULFURTRANSFERASE, MERCAPTOPYRUVATE SULFURTRANSFERASE 2, MST2, ST2, STR2, SULFURTRANSFERASE 2, rhodanese homologue 2
Chromosome Location
Chr.1: 5619545 - 5622674 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035979.1 NP_001031056.1 5 - 6 586 105
NM_001198083.1 NP_001185012.1 6 - 7 659 105
NM_001332226.1 NP_001322506.1 5 - 6 570 105
NM_001332227.1 NP_001322507.1 5 - 6 580 105
NM_101511.6 NP_563998.1 5 - 6 630 105
NM_179344.2 NP_849675.1 5 - 6 570 105
GenBank mRNA AB032865.1 - 5 - 6 492 105
AJ010500.1 - 5 - 6 500 105
AK229282.1 - 5 - 6 589 105
AY084558.1 - 5 - 6 570 105
BT024876.1 - 4 - 5 427 105

View Full Product Details