Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FUG1
- Entrez Gene ID
-
838293
- Gene Name
- Translation initiation factor 2, small GTP-binding protein
- Gene Aliases
-
AT1G17220, F20D23.8, F20D23_8, fu-gaeri1
- Chromosome Location
-
Chr.1: 5885007 - 5890500 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101583.4 |
NP_173165.1 |
1 - 2 |
1916 |
106 |
View Full Product Details