Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AlaAT1
- Entrez Gene ID
-
838301
- Gene Name
- alanine aminotransferas
- Gene Aliases
-
ALANINE AMINOTRANSFERAS, AT1G17290, T13M22.3, T13M22_3, alanine aminotransferas
- Chromosome Location
-
Chr.1: 5922332 - 5926400 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101591.6 |
NP_173173.3 |
10 - 11 |
1527 |
73 |
View Full Product Details