Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP1B
Entrez Gene ID
838309
Gene Name
oligouridylate binding protein 1B
Gene Aliases
AT1G17370, F28G4.17, oligouridylate binding protein 1B
Chromosome Location
Chr.1: 5951542 - 5955143 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123829.1 NP_001117301.1 4 - 5 562 70
NM_101598.3 NP_564018.1 4 - 5 668 70
GenBank mRNA AY085705.1 - 4 - 5 470 70
BT026477.1 - 4 - 5 350 70

View Full Product Details