Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUP9
Entrez Gene ID
838402
Gene Name
purine permease 9
Gene Aliases
AT1G18220, ATPUP9, T10F20.24, purine permease 9
Chromosome Location
Chr.1: 6269670 - 6269807 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101682.1 NP_173260.1 1 - 1 82 102

View Full Product Details