Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MAML-4
- Entrez Gene ID
-
838431
- Gene Name
- methylthioalkylmalate synthase-like 4
- Gene Aliases
-
AT1G18500, F15H18.3, F15H18_3, IPMS1, ISOPROPYLMALATE SYNTHASE 1, methylthioalkylmalate synthase-like 4
- Chromosome Location
-
Chr.1: 6368966 - 6373080 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101708.3 |
NP_173285.2 |
7 - 8 |
1703 |
77 |
View Full Product Details