Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBL12
Entrez Gene ID
838441
Gene Name
RHOMBOID-like protein 12
Gene Aliases
ARABIDOPSIS RHOMBOID-LIKE PROTEIN 12, AT1G18600, ATRBL12, AtPARL, F25I16.6, F25I16_6, PARL, RHOMBOID-like protein 12, presenilins-associated rhomboid-like protein
Chromosome Location
Chr.1: 6400282 - 6402794 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101718.5 NP_564058.1 7 - 8 1279 91
GenBank mRNA AF370504.1 - 7 - 8 1083 91
AY081685.1 - 7 - 8 914 91
AY086276.1 - 7 - 8 932 91

View Full Product Details