Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSP
Entrez Gene ID
838445
Gene Name
3-phosphoserine phosphatase
Gene Aliases
3-PHOSPHOSERINE PHOSPHATASE, 3-phosphoserine phosphatase, AT1G18640, PSP1, phosphoserine phosphatase 1
Chromosome Location
Chr.1: 6416169 - 6418693 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332354.1 NP_001320464.1 6 - 7 995 75
NM_202129.3 NP_973858.1 7 - 8 913 75
GenBank mRNA AY065351.1 - 7 - 8 904 75
AY087385.1 - 7 - 8 865 75
AY096687.1 - 6 - 7 767 75

View Full Product Details