Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCB2;3
Entrez Gene ID
838650
Gene Name
Cyclin B2;3
Gene Aliases
AT1G20610, Cyclin B2;3, F5M15.6
Chromosome Location
Chr.1: 7134659 - 7137828 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101912.3 NP_173485.1 7 - 8 873 147
GenBank mRNA BT029740.1 - 6 - 7 566 147

View Full Product Details