Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUC-L1
Entrez Gene ID
841314
Gene Name
nucleolin like 1
Gene Aliases
AT1G48920, ATNUC-L1, F27K7.6, F27K7_6, NUC1, PARALLEL 1, PARL1, nucleolin 1, nucleolin like 1
Chromosome Location
Chr.1: 18098081 - 18101732 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103785.4 NP_175322.1 2 - 3 170 104
GenBank mRNA BT005793.1 - 2 - 3 65 104

View Full Product Details