Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QSOX2
Entrez Gene ID
814654
Gene Name
quiescin-sulfhydryl oxidase 2
Gene Aliases
AT2G01270, AtQSOX2, F10A8.15, F10A8_15, quiescin-sulfhydryl oxidase 2
Chromosome Location
Chr.2: 139309 - 142560 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335047.1 NP_001324094.1 2 - 3 721 97
NM_001335048.1 NP_001324093.1 3 - 4 604 97
NM_126188.4 NP_565258.1 3 - 4 456 97
GenBank mRNA AY090364.1 - 3 - 4 430 97
AY125539.1 - 3 - 4 308 97
BT000880.1 - 3 - 4 308 97

View Full Product Details