Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STL2P
Entrez Gene ID
814675
Gene Name
SEC12P-like 2 protein
Gene Aliases
AT2G01470, ATSEC12, F2I9.9, F2I9_9, SEC12P-like 2 protein
Chromosome Location
Chr.2: 211913 - 214473 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126208.4 NP_178256.1 9 - 10 1130 66
GenBank mRNA AY074332.1 - 9 - 10 1121 66
AY123030.1 - 9 - 10 1092 66

View Full Product Details