Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATMAP65-6
Entrez Gene ID
814722
Gene Name
Microtubule associated protein (MAP65/ASE1) family protein
Gene Aliases
AT2G01910, MAP65-6, T23K3.10, T23K3_10
Chromosome Location
Chr.2: 416021 - 420661 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035868.2 NP_001030945.1 2 - 3 262 94
NM_126252.4 NP_178300.3 2 - 3 1449 94
GenBank mRNA AK317525.1 - 2 - 3 560 94

View Full Product Details