Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSTZ1
Entrez Gene ID
814770
Gene Name
glutathione S-transferase zeta 1
Gene Aliases
AT2G02390, ATGSTZ1, AtMAAI, GLUTATHIONE S-TRANSFERASE 18, GST18, MAAI, T16F16.18, T16F16_18, glutathione S-transferase zeta 1, maleylacetoacetate isomerase
Chromosome Location
Chr.2: 628730 - 631165 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126296.5 NP_178344.1 2 - 3 373 81
NM_179595.5 NP_849926.2 2 - 3 204 81
NM_201671.3 NP_973400.1 2 - 3 226 81
GenBank mRNA AF288182.1 - 1 - 2 88 81
AK226342.1 - 2 - 3 137 81
AY052332.1 - 2 - 3 141 81
AY061901.1 - 1 - 2 88 81
AY208155.1 - 2 - 3 139 81

View Full Product Details