Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AL6
Entrez Gene ID
814776
Gene Name
alfin-like 6
Gene Aliases
AT2G02470, T16F16.26, T16F16_26, alfin-like 6
Chromosome Location
Chr.2: 652564 - 654985 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202573.2 NP_001189502.1 1 - 2 361 75
NM_001335119.1 NP_001323925.1 1 - 2 349 75
NM_126302.3 NP_178351.1 1 - 2 349 75
GenBank mRNA BT003905.1 - 1 - 2 203 75
BT005004.1 - 1 - 2 88 75

View Full Product Details