Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUT2
Entrez Gene ID
814817
Gene Name
sucrose transporter 2
Gene Aliases
ARABIDOPSIS THALIANA SUCROSE TRANSPORTER 3, AT2G02860, ATSUC3, ATSUT2, SUC3, SUCROSE TRANSPORTER 3, T17M13.3, T17M13_3, sucrose transporter 2
Chromosome Location
Chr.2: 828279 - 832619 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126341.3 NP_178389.1 11 - 12 1829 97
NM_201675.2 NP_973404.1 11 - 12 1804 97
GenBank mRNA AJ289165.1 - 11 - 12 1506 97
AK226970.1 - 11 - 12 1633 97

View Full Product Details