Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNS1
Entrez Gene ID
814828
Gene Name
ribonuclease 1
Gene Aliases
AT2G02990, ATRNS1, RIBONUCLEASE 1, T17M13.16, T17M13_16, ribonuclease 1
Chromosome Location
Chr.2: 873506 - 874886 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126351.3 NP_178399.1 3 - 4 683 110
GenBank mRNA AY072413.1 - 3 - 4 518 110
AY114710.1 - 3 - 4 475 110
DQ446504.1 - 3 - 4 475 110
U05206.1 - 3 - 4 562 110

View Full Product Details