Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGP11
Entrez Gene ID
821093
Gene Name
arabinogalactan protein 11
Gene Aliases
ARABINOGALACTAN PROTEIN 11, AT3G01700, ATAGP11, F4P13.24, F4P13_24, arabinogalactan protein 11
Chromosome Location
Chr.3: 258879 - 259631 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111036.3 NP_566146.1 1 - 1 540 88
GenBank mRNA AK118478.1 - 1 - 1 540 88

View Full Product Details