Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SOX
Entrez Gene ID
820118
Gene Name
sulfite oxidase
Gene Aliases
AT-SO, AT3G01910, AtSO, F28J7.38, F28J7_38, SULFITE OXIDASE, sulfite oxidase
Chromosome Location
Chr.3: 314693 - 317431 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084620.1 NP_001078089.1 1 - 2 112 94
NM_111057.4 NP_186840.1 1 - 2 231 94
GenBank mRNA AB071965.1 - 1 - 2 231 94
AF360247.1 - 1 - 2 112 94
AK226455.1 - 1 - 2 113 94
AY133863.1 - 1 - 2 74 94

View Full Product Details