Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKOR
Entrez Gene ID
821052
Gene Name
STELAR K+ outward rectifier
Gene Aliases
AT3G02850, F13E7.21, F13E7_21, STELAR K+ outward rectifier
Chromosome Location
Chr.3: 619678 - 623739 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337437.1 NP_001325630.1 6 - 7 1509 94
NM_001337438.1 NP_001325629.1 6 - 7 1328 94
NM_111153.4 NP_186934.1 6 - 7 1509 94

View Full Product Details